Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178676.4 | NP_848791.2 | 10 | 1359 | ||
GenBank mRNA | AK032057.1 | - | 10 | 1345 | ||
AK046218.1 | - | 10 | 1335 | |||
AK087561.1 | - | 10 | 1333 | |||
AK169947.1 | - | 5 | 769 | |||
AK170974.1 | - | 5 | 768 | |||
AY376710.1 | - | 10 | 1327 | |||
AY714060.1 | - | 10 | 1330 | |||
BC079871.1 | - | 10 | 1330 | 6831925 |