Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrtm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178678.4 | NP_848793.1 | 2 | 709 | ||
GenBank mRNA | AK032485.1 | - | 2 | 726 | ||
AK035773.1 | - | 2 | 706 | |||
AK035858.1 | - | 2 | 712 | |||
AK035882.1 | - | 2 | 710 | |||
AK036180.1 | - | 2 | 729 | |||
AK036366.1 | - | 2 | 703 | |||
AK163753.1 | - | 2 | 702 | |||
AY182029.1 | - | 1 | 167 | |||
BC094910.1 | - | 2 | 196 | 30044805 | ||
BC113178.1 | - | 2 | 383 | 40060681 | ||
BC113784.1 | - | 2 | 383 | 40060682 |