Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178693.4 | NP_848808.1 | 7 | 1035 | ||
GenBank mRNA | AK039422.1 | - | 7 | 1036 | ||
AK039676.1 | - | 7 | 1036 | |||
AK043574.1 | - | 7 | 1011 | |||
AK050459.1 | - | 7 | 1045 | |||
AK163270.1 | - | 1881 | ||||
BC039570.1 | - | 2 | 683 |