Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178695.5 | NP_848810.3 | 4, 5 | 636 | ||
GenBank mRNA | AK035530.1 | - | 4, 5 | 461 | ||
AK037087.1 | - | 4, 5 | 556 | |||
AK038703.1 | - | 4, 5 | 636 | |||
AK166405.1 | - | 3, 4 | 597 | |||
BC099390.1 | - | 4, 5 | 553 | 30715383 | ||
BC115470.2 | - | 4, 5 | 382 | 40100436 | ||
BC151011.1 | - | 4, 5 | 566 | 9087924 | ||
BC151019.1 | - | 4, 5 | 566 | 9087932 |