Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc8d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122768.1 | NP_001116240.1 | 3 | 572 | ||
NM_178701.3 | NP_848816.3 | 3 | 562 | |||
GenBank mRNA | AK046249.1 | - | 3 | 571 | ||
AK050400.1 | - | 3 | 532 | |||
AK080722.1 | - | 3 | 506 | |||
AK137459.1 | - | 3 | 573 | |||
AK197495.1 | - | 1 | 65 | |||
BC037717.1 | - | 3 | 543 | 4239389 |