Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luzp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178705.5 | NP_848820.4 | 7 | 1211 | ||
GenBank mRNA | AK032204.1 | - | 7 | 868 | ||
AK034077.1 | - | 7 | 829 | |||
AK034240.1 | - | 7 | 863 | |||
AK082126.1 | - | 7 | 959 | |||
AY164459.1 | - | 7 | 1208 | |||
BC075646.1 | - | 7 | 841 | 30620475 |