Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161410.1 | NP_001154882.1 | 2 | 187 | ||
NM_001161411.1 | NP_001154883.1 | 2 | 413 | |||
NM_178811.4 | NP_848926.3 | 2 | 413 | |||
GenBank mRNA | AK031763.1 | - | 2 | 337 | ||
AK052245.1 | - | 2 | 223 | |||
AK084171.1 | - | 2 | 259 | |||
AK086217.1 | - | 2 | 343 | |||
AK132166.1 | - | 2 | 199 | |||
AK147985.1 | - | 2 | 187 | |||
AK171540.1 | - | 2 | 223 | |||
BC024077.1 | - | 2 | 356 | 5097580 | ||
BC030887.1 | - | 2 | 181 | 4483611 |