Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161410.1 | NP_001154882.1 | 5 | 1634 | ||
NM_001161411.1 | NP_001154883.1 | 5 | 1860 | |||
NM_178811.4 | NP_848926.3 | 5 | 1860 | |||
GenBank mRNA | AK031763.1 | - | 5 | 1784 | ||
AK049172.1 | - | 4 | 894 | |||
AK052245.1 | - | 5 | 1670 | |||
AK078174.1 | - | 4 | 435 | |||
AK086217.1 | - | 5 | 1790 | |||
AK132166.1 | - | 5 | 1646 | |||
AK147985.1 | - | 5 | 1634 | |||
AK189551.1 | - | 2 | 173 | |||
BC024077.1 | - | 5 | 1803 | 5097580 | ||
BC030887.1 | - | 5 | 1628 | 4483611 |