Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfesd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131068.1 | NP_001124540.1 | 4 | 875 | ||
NM_001131069.1 | NP_001124541.1 | 4 | 338 | |||
NM_178916.5 | NP_849247.3 | 4 | 1325 | |||
GenBank mRNA | AK029602.1 | - | 4 | 1151 | ||
AK040255.1 | - | 4 | 830 | |||
AK083705.1 | - | 4 | 1326 | |||
BC030381.1 | - | 4 | 950 | 5375753 | ||
CD769672.1 | - | 4 | 342 | |||
CN839985.1 | - | 4 | 348 |