Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286040.1 | NP_001272969.1 | 2 | 368 | ||
NM_001286041.1 | NP_001272970.1 | 2 | 368 | |||
NM_181413.4 | NP_852078.1 | 2 | 368 | |||
GenBank mRNA | AK129094.1 | - | 2 | 302 | ||
AK147273.1 | - | 2 | 303 | |||
BC027740.1 | - | 2 | 354 | |||
BC050847.1 | - | 2 | 334 | 6507130 |