Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc85a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166661.2 | NP_001160133.1 | 3 | 2013 | ||
NM_181577.5 | NP_853555.2 | 3 | 2013 | |||
GenBank mRNA | AK045568.1 | - | 3 | 1986 | ||
AK173298.1 | - | 3 | 2030 | |||
BC020949.1 | - | 2 | 976 | |||
BC066065.1 | - | 4 | 1452 | 6850852 |