Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183017.2 | NP_898838.2 | 14 | 2112 | ||
GenBank mRNA | AK084879.1 | - | 14 | 2081 | ||
AK134345.1 | - | 14 | 2082 | |||
AK143496.1 | - | 13 | 1945 | |||
AK150116.1 | - | 14 | 2078 | |||
AK179941.1 | - | 1 | 41 | |||
BC055368.1 | - | 14 | 2077 | 30055801 |