Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198604.2 | NP_941006.2 | 15 | 2001 | ||
GenBank mRNA | AK153619.1 | - | 15 | 2291 | ||
BC026778.1 | - | 15 | 2004 | 5133546 |