Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198958.2 | NP_945196.2 | 5, 6 | 544 | ||
GenBank mRNA | AY182377.1 | - | 5, 6 | 544 | ||
BC106862.2 | - | 5, 6 | 544 | 40047310 |