Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005911.5 | NP_005902.1 | 2 | 462 | ||
GenBank mRNA | AK291126.1 | - | 2 | 270 | ||
AK293841.1 | - | 2 | 272 | |||
AK297758.1 | - | 2 | 165 | |||
AK308591.1 | - | 2 | 165 | |||
AK308758.1 | - | 2 | 130 | |||
AK309338.1 | - | 2 | 270 | |||
AK316411.1 | - | 2 | 420 | |||
BC001686.2 | - | 2 | 259 | 3010820 | ||
BC001854.2 | - | 2 | 259 | 3010820 | ||
BI549760.1 | - | 2 | 281 | |||
DC322518.1 | - | 2 | 303 | |||
DQ083239.1 | - | 2 | 198 | |||
X68836.1 | - | 2 | 217 |