Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023223.2 | NP_075712.2 | 6 | 1001 | ||
GenBank mRNA | AB045313.1 | - | 6 | 905 | ||
AF312208.1 | - | 6 | 898 | |||
AK011131.1 | - | 4 | 388 | |||
AK011723.1 | - | 6 | 1001 | |||
AK029424.1 | - | 6 | 953 | |||
AK075998.1 | - | 6 | 949 | |||
AK076030.1 | - | 6 | 953 | |||
AK083459.1 | - | 6 | 830 | |||
AK160204.1 | - | 6 | 1013 | |||
AK168650.1 | - | 6 | 953 | |||
AK188208.1 | - | 2 | 236 | |||
AK206746.1 | - | 2 | 237 | |||
AK206928.1 | - | 2 | 236 | |||
AK207342.1 | - | 2 | 236 | |||
AK207468.1 | - | 2 | 236 | |||
AK208541.1 | - | 2 | 236 | |||
AK208620.1 | - | 2 | 236 | |||
AK209126.1 | - | 2 | 236 | |||
AK209399.1 | - | 2 | 239 | |||
AK210837.1 | - | 2 | 236 | |||
AK211703.1 | - | 2 | 236 | |||
AK213222.1 | - | 2 | 236 | |||
AK215738.1 | - | 2 | 236 | |||
AK215892.1 | - | 2 | 236 | |||
AK217035.1 | - | 2 | 236 | |||
AK219314.1 | - | 2 | 237 | |||
AK219440.1 | - | 2 | 236 | |||
AK219805.1 | - | 2 | 237 | |||
AK219889.1 | - | 2 | 236 | |||
BC003215.1 | - | 6 | 938 | 3157842 |