Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ypel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291047.1 | NP_001277976.1 | 6 | 886 | ||
NM_001291053.1 | NP_001277982.1 | 4 | 480 | |||
NM_023249.6 | NP_075738.1 | 5 | 760 | |||
GenBank mRNA | AF190624.1 | - | 5 | 676 | ||
AK005955.1 | - | 5 | 596 | |||
AK006505.1 | - | 5 | 571 | |||
AK006858.1 | - | 5 | 648 | |||
AK011452.1 | - | 5 | 691 | |||
AK014931.1 | - | 5 | 586 | |||
AK015752.1 | - | 4 | 597 | |||
AK018910.1 | - | 5 | 571 | |||
AK019819.1 | - | 5 | 591 | |||
AK133401.1 | - | 5 | 674 | |||
AK139453.1 | - | 5 | 668 | |||
BC049737.1 | - | 4 | 390 | 6771362 | ||
BQ573264.1 | - | 4 | 446 |