Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr7a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025337.3 | NP_079613.3 | 7 | 1208 | ||
GenBank mRNA | AJ271800.1 | - | 7 | 1125 | ||
AJ271801.1 | - | 7 | 1125 | |||
AK002664.1 | - | 7 | 1188 | |||
AK003915.1 | - | 7 | 1196 | |||
AK143203.1 | - | 7 | 1207 | |||
AK193156.1 | - | 1 | 165 | |||
BC031857.1 | - | 7 | 1185 | 5124841 |