Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Manba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027288.3 | NP_081564.3 | 2 | 279 | ||
GenBank mRNA | AF306557.1 | - | 2 | 283 | ||
AK036505.1 | - | 2 | 277 | |||
AK047057.1 | - | 2 | 272 | |||
AK142165.1 | - | 2 | 244 | |||
AK154172.1 | - | 2 | 257 | |||
AK166022.1 | - | 2 | 261 | |||
AK166340.1 | - | 2 | 277 | |||
AK170290.1 | - | 2 | 276 | |||
AK171416.1 | - | 2 | 266 | |||
BC031409.1 | - | 2 | 222 | 4952710 |