Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Manba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027288.3 | NP_081564.3 | 6 | 891 | ||
GenBank mRNA | AF306557.1 | - | 6 | 895 | ||
AK036505.1 | - | 7 | 991 | |||
AK047057.1 | - | 6 | 884 | |||
AK142165.1 | - | 6 | 856 | |||
AK154172.1 | - | 6 | 872 | |||
AK166022.1 | - | 6 | 873 | |||
AK166340.1 | - | 6 | 889 | |||
AK170290.1 | - | 6 | 888 | |||
BC031409.1 | - | 6 | 834 | 4952710 |