Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col16a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028266.5 | NP_082542.3 | 70 | 5054 | ||
GenBank mRNA | AK012212.1 | - | 3 | 686 | ||
AK031163.1 | - | 61 | 3919 | |||
AK040971.1 | - | 70 | 5007 | |||
BC018293.1 | - | 1 | 141 | |||
BC023940.1 | - | 24 | 1763 | |||
BC027766.1 | - | 70 | 4989 | 5346676 |