Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163529.1 | NP_001157001.1 | 16 | 2168 | ||
NM_033615.2 | NP_291093.2 | 16 | 2168 | |||
GenBank mRNA | AB059632.1 | - | 16 | 2168 | ||
AB059633.1 | - | 16 | 2168 | |||
AF386072.1 | - | 12 | 1535 | |||
AF472524.1 | - | 16 | 1900 | |||
AK048367.1 | - | 12 | 1435 | |||
AK053825.1 | - | 14 | 2426 | |||
AK138517.1 | - | 2 | 512 | |||
AY054984.1 | - | 16 | 1868 | |||
AY382193.1 | - | 16 | 1899 | |||
AY382194.1 | - | 16 | 1899 | |||
AY953137.1 | - | 11 | 1443 | |||
BC138139.1 | - | 16 | 1891 | 8861158 |