Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053246.3 | NP_444476.1 | 5 | 721 | ||
GenBank mRNA | AF418207.1 | - | 4 | 605 | ||
AK032352.1 | - | 5 | 711 | |||
AK043670.1 | - | 4 | 633 | |||
AK077612.1 | - | 5 | 713 | |||
AK139363.1 | - | 4 | 609 | |||
AK142621.1 | - | 4 | 628 | |||
AK156485.1 | - | 5 | 721 | |||
AK192791.1 | - | 2 | 205 | |||
BC004705.1 | - | 5 | 682 | 3499419 |