Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgb3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289643.1 | NP_001276572.1 | 3 | 508 | ||
NM_001289644.1 | NP_001276573.1 | 3 | 442 | |||
GenBank mRNA | AF274959.1 | - | 3 | 416 | ||
AF274960.1 | - | 3 | 482 | |||
AF274961.1 | - | 2 | 746 | |||
AF439546.1 | - | 3 | 408 | |||
AK136373.1 | - | 3 | 420 | |||
AK136396.1 | - | 3 | 419 | |||
BC061046.1 | - | 416 | 30301032 |