Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054066.4 | NP_473407.2 | 4 | 405 | ||
GenBank mRNA | AF435950.1 | - | 4 | 410 | ||
AK005949.1 | - | 4 | 414 | |||
AK006672.1 | - | 4 | 450 | |||
BC106767.1 | - | 4 | 400 | 40039566 | ||
BC106768.1 | - | 4 | 403 | 40039567 |