Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afg3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054070.3 | NP_473411.2 | 8 | 1034 | ||
GenBank mRNA | AF329695.1 | - | 8 | 975 | ||
AK006708.1 | - | 2 | 325 | |||
AK159647.1 | - | 8 | 998 | |||
AK167964.1 | - | 8 | 1009 | |||
AK168244.1 | - | 8 | 1008 | |||
BC056978.1 | - | 8 | 993 | 6821746 | ||
BC060064.1 | - | 8 | 993 |