Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prelp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054077.4 | NP_473418.3 | 2 | 247 | ||
GenBank mRNA | AK037150.1 | - | 2 | 225 | ||
AK047480.1 | - | 2 | 248 | |||
AK205311.1 | - | 2 | 137 | |||
BC019775.1 | - | 2 | 223 | 4165772 |