Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080465.2 | NP_536713.1 | 6 | 1307 | ||
GenBank mRNA | AF357240.1 | - | 6 | 1307 | ||
AF533008.1 | - | 6 | 1307 | |||
AK033158.1 | - | 9 | 2811 | |||
AK050390.1 | - | 6 | 1563 | |||
AK149082.1 | - | 6 | 1995 | |||
AY123778.1 | - | 6 | 1382 | |||
BC125475.1 | - | 6 | 1333 | 40129990 | ||
BC137656.1 | - | 6 | 1333 | 8860675 | ||
EU478850.1 | - | 5 | 887 |