Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubox5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001255993.1 | NP_001242922.1 | 7 | 2868 | ||
NM_001255994.1 | NP_001242923.1 | 6 | 2721 | |||
NM_080562.5 | NP_542129.2 | 6 | 2726 | |||
GenBank mRNA | AK034114.1 | - | 6 | 2675 |