Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf144a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081977.2 | NP_001075446.1 | 10 | 1712 | ||
NM_080563.4 | NP_542130.1 | 9 | 1638 | |||
GenBank mRNA | AK038586.1 | - | 9 | 1530 | ||
AK133544.1 | - | 7 | 1252 | |||
AK172904.1 | - | 8 | 1474 | |||
BC030187.1 | - | 9 | 1603 | 5345434 |