Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caskin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080643.2 | NP_542374.2 | 6 | 948 | ||
GenBank mRNA | AF451978.1 | - | 5 | 667 | ||
AK129297.1 | - | 6 | 986 | |||
BC053083.1 | - | 6 | 948 | 5704642 |