Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APBB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006051.3 | NP_006042.3 | 6 | 895 | ||
NM_133172.2 | NP_573418.2 | 6 | 895 | |||
NM_133173.2 | NP_573419.2 | 6 | 895 | |||
NM_133174.2 | NP_573420.2 | 6 | 895 | |||
GenBank mRNA | AB018247.1 | - | 6 | 587 | ||
AB049618.1 | - | 6 | 623 | |||
AB061224.1 | - | 6 | 536 | |||
AB079687.1 | - | 6 | 536 | |||
AB094990.1 | - | 6 | 536 | |||
AB209839.1 | - | 6 | 623 | |||
AF224708.1 | - | 6 | 556 | |||
AF224709.1 | - | 6 | 556 | |||
AF224710.1 | - | 6 | 556 | |||
AF224711.1 | - | 6 | 556 | |||
AK074885.1 | - | 3 | 430 | |||
AK075280.1 | - | 6 | 858 | |||
AK125244.1 | - | 5 | 1156 | |||
AK304824.1 | - | 4 | 524 | |||
BC013158.1 | - | 6 | 895 | 3870148 | ||
BC125187.1 | - | 6 | 887 | 40122344 |