Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adrbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290818.1 | NP_001277747.1 | 21 | 2946 | ||
NM_130863.2 | NP_570933.1 | 22 | 3018 | |||
GenBank mRNA | AK153543.1 | - | 3 | 970 | ||
AK155691.1 | - | 21 | 2947 | |||
BC033272.1 | - | 18 | 3094 | |||
BC053922.1 | - | 22 | 2751 | 6309555 |