Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cstf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290398.1 | NP_001277327.1 | 14 | 3070 | ||
NM_133196.6 | NP_573459.1 | 14 | 3148 | |||
GenBank mRNA | AK047255.1 | - | 2 | 2054 | ||
AK049971.1 | - | 14 | 3061 | |||
AK088260.1 | - | 14 | 2976 | |||
AK147438.1 | - | 12 | 6142 |