Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134050.4 | NP_598811.3 | 7 | 2417 | ||
GenBank mRNA | AK158637.1 | - | 7 | 2339 | ||
AK158768.1 | - | 7 | 2371 | |||
BC013790.1 | - | 7 | 1837 | 2655151 | ||
BC023739.1 | - | 1 | 675 | |||
BC027769.1 | - | 7 | 2413 | 5345297 |