Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptssa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134054.2 | NP_598815.2 | 2 | 680 | ||
GenBank mRNA | AK003171.1 | - | 2 | 617 | ||
AK003275.1 | - | 2 | 621 | |||
AK003754.1 | - | 1 | 76 | |||
AK008904.1 | - | 2 | 637 | |||
AK018380.1 | - | 2 | 657 | |||
AK137543.1 | - | 2 | 655 | |||
BC022674.1 | - | 2 | 606 | 4236922 |