Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam19a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252310.1 | NP_001239239.1 | 4 | 1345 | ||
NM_134096.2 | NP_598857.1 | 4 | 1297 | |||
GenBank mRNA | AK090194.1 | - | 4 | 1172 | ||
AK090307.1 | - | 4 | 1348 | |||
BC015306.1 | - | 4 | 1298 | 4223181 |