Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134136.3 | NP_598897.2 | 18 | 3085 | ||
GenBank mRNA | AK031096.1 | - | 18 | 3035 | ||
AK031347.1 | - | 18 | 3016 | |||
AK089046.1 | - | 18 | 3054 | |||
AY267463.1 | - | 18 | 3015 | |||
BC056348.1 | - | 18 | 3037 | 5721407 |