Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134155.1 | NP_598916.1 | 4 | 353 | ||
GenBank mRNA | AF233580.1 | - | 4 | 364 | ||
AK076729.1 | - | 4 | 361 | |||
BC003485.1 | - | 4 | 371 | 2811430 | ||
BC016108.1 | - | 4 | 353 | 4921658 | ||
BC038380.1 | - | 2592 |