Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fdps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253751.1 | NP_001240680.1 | 3, 4 | 490 | ||
NM_134469.4 | NP_608219.1 | 2, 3 | 274 | |||
GenBank mRNA | AF309508.1 | - | 1, 2 | 179 | ||
AK088601.1 | - | 2, 3 | 194 | |||
AK140881.1 | - | 2, 3 | 266 | |||
AK151742.1 | - | 2, 3 | 203 | |||
AK166026.1 | - | 2, 3 | 193 | |||
AK168008.1 | - | 2, 3 | 193 | |||
BC048497.1 | - | 2, 3 | 252 | 6531586 | ||
BC087886.1 | - | 2, 3 | 197 | 6595030 | ||
CJ082322.1 | - | 2, 3 | 274 | |||
CT010365.1 | - | 1, 2 | 146 |