Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138646.3 | NP_619587.3 | 6 | 996 | ||
GenBank mRNA | AK081489.1 | - | 6 | 956 | ||
AK129418.1 | - | 5 | 883 | |||
AK147954.1 | - | 5 | 1439 | |||
AK171871.1 | - | 6 | 965 | |||
AK187573.1 | - | 3 | 229 | |||
AY043415.1 | - | 5 | 516 | |||
BC063055.1 | - | 5 | 688 | 5701206 |