Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138669.1 | NP_619610.1 | 11 | 1224 | ||
GenBank mRNA | AK075920.1 | - | 11 | 1200 | ||
AK150277.1 | - | 11 | 1240 | |||
AK151786.1 | - | 11 | 1202 | |||
AK152824.1 | - | 11 | 1240 | |||
AK153359.1 | - | 11 | 1206 | |||
AK157855.1 | - | 11 | 1200 | |||
AK167107.1 | - | 11 | 1262 | |||
AK168319.1 | - | 11 | 1204 | |||
AK169350.1 | - | 11 | 1204 | |||
AK169815.1 | - | 2630 | ||||
BC008132.1 | - | 11 | 1224 | 3498954 | ||
BC012862.1 | - | 11 | 1212 | 3585696 |