Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138669.1 | NP_619610.1 | 11 | 1172 | ||
GenBank mRNA | AK075920.1 | - | 11 | 1148 | ||
AK150277.1 | - | 11 | 1188 | |||
AK151786.1 | - | 11 | 1150 | |||
AK152824.1 | - | 11 | 1188 | |||
AK153359.1 | - | 11 | 1154 | |||
AK157855.1 | - | 11 | 1148 | |||
AK167107.1 | - | 11 | 1210 | |||
AK168319.1 | - | 11 | 1152 | |||
AK169350.1 | - | 11 | 1152 | |||
AK169815.1 | - | 2578 | ||||
AK183369.1 | - | 2 | 160 | |||
BC008132.1 | - | 11 | 1172 | 3498954 | ||
BC012862.1 | - | 11 | 1160 | 3585696 |
Gene Symbol: Gm5580 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC158083.1 | - | 1 | 1148 | 9088173 |
Gene Symbol: Gm8994 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142734.1 | NP_001136206.1 | 2 | 1257 | ||
GenBank mRNA | AK139713.1 | - | 2 | 1259 |