Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Luc7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170848.1 | NP_001164319.1 | 9 | 2044 | ||
NM_138680.2 | NP_619621.2 | 10 | 2074 | |||
GenBank mRNA | AF318301.1 | - | 10 | 1999 | ||
AK134191.1 | - | 9 | 1958 | |||
AK147889.1 | - | 12 | 2214 | |||
AK153527.1 | - | 13 | 2302 | |||
BC020058.1 | - | 11 | 1799 | 4009362 | ||
BC056354.1 | - | 10 | 2072 | 6811460 | ||
BC056970.1 | - | 10 | 2072 | 6811460 |