Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138955.3 | NP_620405.3 | 15 | 2826 | ||
GenBank mRNA | AF378330.1 | - | 14 | 2658 | ||
AF411084.1 | - | 15 | 2704 | |||
AF425077.1 | - | 15 | 2705 | |||
AJ426047.2 | - | 15 | 2705 | |||
AK018283.1 | - | 1 | 626 | |||
AK043147.1 | - | 15 | 2827 | |||
AK157317.1 | - | 15 | 3151 | |||
BC016200.1 | - | 15 | 2706 | 4511456 | ||
BC023077.1 | - | 9 | 1914 | |||
BC030301.1 | - | 1 | 1022 | |||
BC059033.1 | - | 6 | 1999 |