Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102420.2 | NP_001095890.1 | 7 | 1700 | ||
NM_001102421.2 | NP_001095891.1 | 6 | 1563 | |||
NM_001278243.1 | NP_001265172.1 | 7 | 1279 | |||
NM_001278244.1 | NP_001265173.1 | 6 | 1142 | |||
NM_001278245.1 | NP_001265174.1 | 7 | 1276 | |||
NM_006007.3 | NP_005998.1 | 6 | 1560 | |||
GenBank mRNA | AF062346.1 | - | 6 | 1283 | ||
AF062347.1 | - | 6 | 1138 | |||
AK025782.1 | - | 7 | 1163 | |||
AK290849.1 | - | 6 | 1283 | |||
AK307376.1 | - | 4 | 984 | |||
BC073131.1 | - | 6 | 1064 | 6137219 | ||
BX648551.1 | - | 1 | 4491 | |||
BX649046.1 | - | 1 | 1088 |