Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAGE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006014.4 | NP_006005.2 | 3 | 859 | ||
GenBank mRNA | BC015744.1 | - | 3 | 859 | 4899044 | |
BC019012.2 | - | 3 | 481 | |||
BC062330.1 | - | 3 | 572 | 5485603 | ||
BQ649305.1 | - | 3 | 526 | |||
X92896.1 | - | 3 | 513 |