Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145615.4 | NP_663590.3 | 7 | 785 | ||
GenBank mRNA | AK028118.1 | - | 7 | 786 | ||
AK032830.1 | - | 7 | 738 | |||
AK133525.1 | - | 7 | 731 | |||
AK167572.1 | - | 7 | 758 | |||
AK168321.1 | - | 7 | 746 | |||
BC003432.1 | - | 7 | 676 | 2646522 | ||
BC096645.1 | - | 7 | 644 | 1245213 |