Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osgin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145950.4 | NP_666062.3 | 3 | 363 | ||
GenBank mRNA | AK048731.1 | - | 3 | 364 | ||
AK053737.1 | - | 3 | 307 | |||
AK148243.1 | - | 3 | 338 | |||
AK169678.1 | - | 3 | 308 | |||
AK186777.1 | - | 2 | 65 | |||
CB574822.1 | - | 3 | 319 |