Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D15Ertd621e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145959.3 | NP_666071.2 | 14 | 1502 | ||
GenBank mRNA | AK122297.1 | - | 14 | 1352 | ||
AK137324.1 | - | 14 | 1503 | |||
BC033609.1 | - | 6 | 584 | 5401763 | ||
BC070426.1 | - | 14 | 1488 | 6405871 | ||
DQ228142.1 | - | 14 | 1281 |